Cyclopropyl(2-methoxy-3,5-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

487590-5G 487590-25G 487590-1G 487590-500MG
77629-948EA 1484.23 USD
77629-948 77635-990 77639-326 77651-308
Cyclopropyl(2-methoxy-3,5-dimethylphenyl)methanone ≥95%
Cyclopropyl(2-methoxy-3,5-dimethylphenyl)methanone

Precaución: For Research Use Only

Formula: 204.265 g/mol
MW: C₁₃H₁₆O₂
Storage Temperature: Ambient
CAS Number: 1466161-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR