Tert-butyl 2-methoxy-3,5-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

487589-5G 487589-25G 487589-1G 487589-500MG
77629-946EA 1484.23 USD
77629-946 77635-988 77643-346 77651-306
Tert-butyl 2-methoxy-3,5-dimethylbenzoate ≥95%
Tert-butyl 2-methoxy-3,5-dimethylbenzoate

Precaución: For Research Use Only

Formula: C₁₄H₂₀O₃
Storage Temperature: Ambient
CAS Number: 3031805-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR