1-(2-Methoxy-3,5-dimethylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

487587-5G 487587-1G 487587-25G 487587-500MG
77633-408EA 1381 USD
77633-408 77639-324 77656-210 77651-304
1-(2-Methoxy-3,5-dimethylphenyl)propan-1-one ≥95%
1-(2-Methoxy-3,5-dimethylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 192.254 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
CAS Number: 5384-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR