2,2,2-Trifluoro-1-(3-(2-methoxyethoxy)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

487567-5G 487567-1G 487567-500MG 487567-25G
77629-942EA 1484.23 USD
77629-942 77639-320 77652-930 77656-208
2,2,2-Trifluoro-1-(3-(2-methoxyethoxy)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(3-(2-methoxyethoxy)phenyl)ethanone

Precaución: For Research Use Only

Formula: 248.198 g/mol
MW: C₁₁H₁₁F₃O₃
Storage Temperature: Ambient
CAS Number: 3037703-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR