2-(3-(2-Methoxyethoxy)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

487563-5G 487563-1G 487563-25G 487563-500MG
77629-940EA 1484.23 USD
77629-940 77639-318 77656-206 77651-302
2-(3-(2-Methoxyethoxy)phenyl)propan-2-ol ≥95%
2-(3-(2-Methoxyethoxy)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 210.27 g/mol
MW: C₁₂H₁₈O₃
Storage Temperature: Ambient
CAS Number: 3037693-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR