(3-(2-Methoxyethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

487556-5G 487556-1G 487556-25G 487556-500MG
77633-406EA 1441 USD
77633-406 77639-316 77656-204 77651-300
(3-(2-Methoxyethoxy)phenyl)(methyl)sulfane ≥95%
(3-(2-Methoxyethoxy)phenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 198.282 g/mol
MW: C₁₀H₁₄O₂S
Storage Temperature: Ambient
CAS Number: 3037682-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR