1-(4-Bromo-1-fluoronaphthalen-2-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

487515-5G 487515-1G 487515-25G 487515-500MG
77629-936EA 1484.23 USD
77629-936 77639-314 77659-754 77651-298
1-(4-Bromo-1-fluoronaphthalen-2-yl)propan-1-one ≥95%
1-(4-Bromo-1-fluoronaphthalen-2-yl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
CAS Number: 3031805-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR