1-([1,1'-Biphenyl]-4-yl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

487504-1G 487504-5G 487504-500MG
77639-312EA 560 USD
77639-312 77656-202 77651-296
1-([1,1'-Biphenyl]-4-yl)-2-methoxyethanone ≥95%
1-([1,1'-Biphenyl]-4-yl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 226.27 g/mol
MW: C₁₅H₁₄O₂
Storage Temperature: Ambient
CAS Number: 96187-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR