Tert-butyl 3-isopropylbenzoate ≥95%

Supplier: AOBChem USA

Warning

487502-5G 487502-25G 487502-1G 487502-500MG
77629-934EA 1484.23 USD
77629-934 77635-984 77639-308 77647-320
Tert-butyl 3-isopropylbenzoate ≥95%
Tert-butyl 3-isopropylbenzoate

Precaución: For Research Use Only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
CAS Number: 3031812-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR