1-(3-Isopropylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

487500-5G 487500-25G 487500-1G 487500-500MG
77633-404EA 1357 USD
77633-404 77656-198 77643-342 77651-294
1-(3-Isopropylphenyl)cyclopentanol ≥95%
1-(3-Isopropylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 204.308 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
CAS Number: 1996988-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR