1-(3-Isopropylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

487499-1G 487499-500MG 487499-5G
77639-306EA 507 USD
77639-306 77652-924 77656-196
1-(3-Isopropylphenyl)cyclobutanol ≥95%
1-(3-Isopropylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
CAS Number: 1999596-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR