1-([1,1'-Biphenyl]-3-yl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

487497-5G 487497-1G 487497-500MG
77659-752EA 2001 USD
77659-752 77643-338 77651-292
1-([1,1'-Biphenyl]-3-yl)-2-methoxyethanone ≥95%
1-([1,1'-Biphenyl]-3-yl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 226.27 g/mol
MW: C₁₅H₁₄O₂
Storage Temperature: Ambient
CAS Number: 1256482-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR