1-(4-Fluoro-[1,1'-biphenyl]-3-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

487492-1G 487492-5G 487492-500MG
77639-304EA 657 USD
77639-304 77656-192 77647-316
1-(4-Fluoro-[1,1'-biphenyl]-3-yl)propan-1-one ≥95%
1-(4-Fluoro-[1,1'-biphenyl]-3-yl)propan-1-one

Precaución: For Research Use Only

Formula: 228.262 g/mol
MW: C₁₅H₁₃FO
Storage Temperature: Ambient
CAS Number: 1553833-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR