Tert-butyl 4-fluoro-[1,1'-biphenyl]-3-carboxylate ≥95%

Supplier: AOBChem USA

Warning

487491-5G 487491-25G 487491-1G 487491-500MG
77629-930EA 1484.23 USD
77629-930 77635-982 77639-302 77651-290
Tert-butyl 4-fluoro-[1,1'-biphenyl]-3-carboxylate ≥95%
Tert-butyl 4-fluoro-[1,1'-biphenyl]-3-carboxylate

Precaución: For Research Use Only

Formula: 272.314 g/mol
MW: C₁₇H₁₇FO₂
Storage Temperature: Ambient
CAS Number: 3037676-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR