2-(4-Fluoro-[1,1'-biphenyl]-3-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

487489-5G 487489-25G 487489-1G 487489-500MG
77629-926EA 1484.23 USD
77629-926 77656-188 77643-336 77647-312
2-(4-Fluoro-[1,1'-biphenyl]-3-yl)propan-2-ol ≥95%
2-(4-Fluoro-[1,1'-biphenyl]-3-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 230.277 g/mol
MW: C₁₅H₁₅FO
Storage Temperature: Ambient
CAS Number: 3037704-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR