(4-Fluoro-[1,1'-biphenyl]-3-yl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

487487-5G 487487-1G 487487-25G 487487-500MG
77633-402EA 1441 USD
77633-402 77639-298 77656-186 77651-288
(4-Fluoro-[1,1'-biphenyl]-3-yl)trimethylsilane ≥95%
(4-Fluoro-[1,1'-biphenyl]-3-yl)trimethylsilane

Precaución: For Research Use Only

Formula: 244.379 g/mol
MW: C₁₅H₁₇FSi
Storage Temperature: Ambient
CAS Number: 3037693-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR