1-(5-Cyclohexyl-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

487475-5G 487475-25G 487475-1G 487475-500MG
77629-920EA 1484.23 USD
77629-920 77659-750 77643-332 77651-284
1-(5-Cyclohexyl-2-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(5-Cyclohexyl-2-fluorophenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: C₁₄H₁₅F₃O
Storage Temperature: Ambient
CAS Number: 3037693-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR