1-(5-Cyclohexyl-2-fluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

487474-5G 487474-1G 487474-25G 487474-500MG
77629-918EA 1484.23 USD
77629-918 77639-294 77656-182 77651-282
1-(5-Cyclohexyl-2-fluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(5-Cyclohexyl-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₁₄H₁₄F₄O
Storage Temperature: Ambient
CAS Number: 3037705-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR