1-(5-Cyclohexyl-2-fluorophenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
487473-5G
487473-25G
487473-1G
487473-500MG
77629-916EA
1484.23
USD
77629-916
77659-748
77643-330
77647-310
1-(5-Cyclohexyl-2-fluorophenyl)ethanone ≥95%
1-(5-Cyclohexyl-2-fluorophenyl)ethanone
Precaución: For Research Use Only
Formula:
C₁₄H₁₇FO Storage Temperature: Ambient |
CAS Number:
3037696-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...