2-Bromo-4-cyclohexyl-1-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

487472-5G 487472-500MG 487472-25G 487472-1G
77629-914EA 1484.23 USD
77629-914 77652-920 77659-746 77643-328
2-Bromo-4-cyclohexyl-1-fluorobenzene ≥95%
2-Bromo-4-cyclohexyl-1-fluorobenzene

Precaución: For Research Use Only

Formula: C₁₂H₁₄BrF
Storage Temperature: Ambient
CAS Number: 3037704-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR