2-(5-Cyclohexyl-2-fluorophenyl)propan-2-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
487471-5G
487471-1G
487471-500MG
487471-25G
77629-912EA
1484.23
USD
77629-912
77639-292
77652-918
77659-744
2-(5-Cyclohexyl-2-fluorophenyl)propan-2-ol ≥95%
2-(5-Cyclohexyl-2-fluorophenyl)propan-2-ol
Precaución: For Research Use Only
Formula:
C₁₅H₂₁FO Storage Temperature: Ambient |
CAS Number:
3037703-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...