(5-Cyclohexyl-2-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

487469-5G 487469-25G 487469-1G 487469-500MG
77629-908EA 1484.23 USD
77629-908 77656-180 77643-326 77647-308
(5-Cyclohexyl-2-fluorophenyl)trimethylsilane ≥97%
(5-Cyclohexyl-2-fluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 250.427 g/mol
MW: C₁₅H₂₃FSi
Storage Temperature: Ambient
CAS Number: 3037703-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR