Neopentyl methanesulfonate ≥95%

Supplier: AOBChem USA

Warning

487439-5G 487439-1G 487439-25G 487439-500MG
77633-400EA 981 USD
77633-400 77639-286 77659-740 77651-278
Neopentyl methanesulfonate ≥95%
Neopentyl methanesulphonate

Precaución: For Research Use Only

Formula: C₆H₁₄O₃S
Storage Temperature: Ambient
CAS Number: 16427-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR