4-(Pyridin-2-ylthio)benzoic acid 95

Supplier: AOBChem USA

Warning

487404-5G 487404-1G 487404-500MG 487404-25G
77633-398EA 780 USD
77633-398 77639-284 77652-916 77656-176
4-(Pyridin-2-ylthio)benzoic acid 95
4-(Pyridin-2-ylthio)benzoic acid

Precaución: For Research Use Only

Formula: C₁₂H₉NO₂S
MW: 231.27 g/mol
Storage Temperature: Ambient
CAS Number: 852952-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR