4-(Pyridin-2-ylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

487402-5G 487402-25G 487402-1G 487402-500MG
77633-396EA 904 USD
77633-396 77656-174 77643-324 77647-304
4-(Pyridin-2-ylthio)benzonitrile ≥97%
4-(Pyridin-2-ylthio)benzonitrile

Precaución: For Research Use Only

Formula: C₁₂H₈N₂S
Storage Temperature: Ambient
CAS Number: 146596-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR