α,α-Dimethyl-2-pyridineethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
487140-5G
487140-1G
487140-500MG
77629-906EA
1484.23
USD
77629-906
77639-282
77647-302
α,α-Dimethyl-2-pyridineethanol ≥95%
α,α-Dimethyl-2-pyridineethanol
Precaución: For Research Use Only
Formula:
C₉H₁₃NO Storage Temperature: Ambient |
CAS Number:
16193-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...