2-[(Methylthio)methyl]pyridine ≥95%

Supplier: AOBChem USA

Warning

487133-1G 487133-500MG 487133-5G
77639-280EA 557 USD
77639-280 77652-914 77659-738
2-[(Methylthio)methyl]pyridine ≥95%
2-[(Methylthio)methyl]pyridine

Precaución: For Research Use Only

Formula: C₇H₉NS
Storage Temperature: Ambient
CAS Number: 3145-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR