6-Fluoro-α,α-dimethyl-2-pyridineethanol ≥95%

Supplier: AOBChem USA

Warning

486915-5G 486915-1G 486915-500MG
77629-904EA 1484.23 USD
77629-904 77643-320 77651-270
6-Fluoro-α,α-dimethyl-2-pyridineethanol ≥95%
6-Fluoro-α,α-dimethyl-2-pyridineethanol

Precaución: For Research Use Only

Formula: C₉H₁₂FNO
Storage Temperature: Ambient
CAS Number: 2354260-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR