1-(6-Fluoro-2-pyridinyl)-2-propanone ≥95%

Supplier: AOBChem USA

Warning

486903-1G 486903-5G 486903-500MG
77639-272EA 528 USD
77639-272 77659-734 77651-264
1-(6-Fluoro-2-pyridinyl)-2-propanone ≥95%
1-(6-Fluoro-2-pyridinyl)-2-propanone

Precaución: For Research Use Only

Formula: 153.15 g/mol
MW: C₈H₈FNO
Storage Temperature: Ambient
CAS Number: 2354041-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR