2-(2-Methyl-1,3-dioxolan-2-yl)thiazole ≥97%

Supplier: AOBChem USA

Warning

486865-5G 486865-1G 486865-500MG
77629-900EA 1484.23 USD
77629-900 77643-316 77647-300
2-(2-Methyl-1,3-dioxolan-2-yl)thiazole ≥97%
2-(2-Methyl-1,3-dioxolan-2-yl)thiazole

Precaución: For Research Use Only

Formula: 171.2168 g/mol
MW: C₇H₉NO₂S
Storage Temperature: Ambient
CAS Number: 24295-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR