2-Bromo-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-[4-(trifluoromethyl)phenyl]benzene

486842-5G 486842-1G 486842-500MG
77656-166EA 2456 USD
77656-166 77643-314 77647-298
2-Bromo-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Bromo-4'-(trifluoromethyl)-1,1'-biphenyl

Precaución: For Research Use Only

Formula: C₁₃H₈BrF₃
MW: 301.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412188
CAS Number: 255837-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR