2,4-Diethoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
48616-5G
48616-1G
48616-25G
77635-078EA
237
USD
77635-078
77639-270
77659-732
2,4-Diethoxybenzoic acid ≥97%
2,4-Diethoxybenzoic acid
Precaución: For Research Use Only
Formula:
C₁₁H₁₄O₄ Storage Temperature: Ambient |
MDL Number:
MFCD03427022 CAS Number: 19074-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...