Tert-butyl 3-(benzyloxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

483705-5G 483705-1G 483705-25G 483705-500MG
77633-390EA 760 USD
77633-390 77639-268 77656-162 77647-296
Tert-butyl 3-(benzyloxy)benzoate ≥97%
Tert-butyl 3-(benzyloxy)benzoate

Precaución: For Research Use Only

Formula: 284.35 g/mol
MW: C₁₈H₂₀O₃
Storage Temperature: Ambient
CAS Number: 378787-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR