1-(2-(Methylthio)-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

483698-5G 483698-25G 483698-1G 483698-500MG
77629-898EA 1484.23 USD
77629-898 77659-730 77643-312 77651-262
1-(2-(Methylthio)-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(2-(Methylthio)-5-(trifluoromethyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 262.291 g/mol
MW: C₁₂H₁₃F₃OS
Storage Temperature: Ambient
CAS Number: 3037696-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR