(2-(Methylthio)-5-(trifluoromethyl)phenyl)(phenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

483686-5G 483686-1G 483686-500MG 483686-25G
77633-388EA 1441 USD
77633-388 77639-266 77652-912 77656-160
(2-(Methylthio)-5-(trifluoromethyl)phenyl)(phenyl)methanone ≥97%
(2-(Methylthio)-5-(trifluoromethyl)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 296.307 g/mol
MW: C₁₅H₁₁F₃OS
Storage Temperature: Ambient
CAS Number: 3037704-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR