2,2,2-Trifluoro-1-(2-(methylthio)-5-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

483682-5G 483682-25G 483682-1G 483682-500MG
77629-894EA 1484.23 USD
77629-894 77656-158 77643-308 77647-294
2,2,2-Trifluoro-1-(2-(methylthio)-5-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-(methylthio)-5-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 288.209 g/mol
MW: C₁₀H₆F₆OS
Storage Temperature: Ambient
CAS Number: 3037692-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR