Tert-butyl 3-(m-tolyl)propiolate ≥97%

Supplier: AOBChem USA

Warning

481534-5G 481534-1G 481534-25G 481534-500MG
77633-382EA 1381 USD
77633-382 77639-264 77659-724 77651-254
Tert-butyl 3-(m-tolyl)propiolate ≥97%
Tert-butyl 3-(m-tolyl)propiolate

Precaución: For Research Use Only

Formula: C₁₄H₁₆O₂
Storage Temperature: Ambient
CAS Number: 2138140-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR