4-Methyl-1-(m-tolyl)pent-1-yn-3-ol ≥95%

Supplier: AOBChem USA

Warning

481533-5G 481533-1G 481533-500MG
77656-150EA 2001 USD
77656-150 77643-294 77651-252
4-Methyl-1-(m-tolyl)pent-1-yn-3-ol ≥95%
4-Methyl-1-(m-tolyl)pent-1-yn-3-ol

Precaución: For Research Use Only

Formula: 188.2655 g/mol
MW: C₁₃H₁₆O
Storage Temperature: Ambient
CAS Number: 1417507-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR