Ethyl 2-oxo-4-(m-tolyl)but-3-ynoate ≥97%

Supplier: AOBChem USA

Warning

481532-5G 481532-25G 481532-1G 481532-500MG
77633-380EA 696 USD
77633-380 77659-722 77643-292 77651-250
Ethyl 2-oxo-4-(m-tolyl)but-3-ynoate ≥97%
Ethyl 2-oxo-4-(m-tolyl)but-3-ynoate

Precaución: For Research Use Only

Formula: 216.2325 g/mol
MW: C₁₃H₁₂O₃
Storage Temperature: Ambient
CAS Number: 1791418-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR