1-(2-Ethoxy-3-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

481456-5G 481456-1G 481456-25G 481456-500MG
77629-884EA 1553.24 USD
77629-884 77639-260 77659-718 77651-246
1-(2-Ethoxy-3-methylphenyl)propan-1-one ≥95%
1-(2-Ethoxy-3-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 192.254 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
CAS Number: 1806381-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR