Ethyl 2-(2-ethoxy-3-methylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

481453-5G 481453-25G 481453-1G 481453-500MG
77629-882EA 1484.23 USD
77629-882 77635-972 77639-258 77651-244
Ethyl 2-(2-ethoxy-3-methylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(2-ethoxy-3-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₃H₁₆O₄
Storage Temperature: Ambient
CAS Number: 3031807-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR