1-(2-Ethoxy-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481450-5G 481450-25G 481450-1G 481450-500MG
77629-880EA 1484.23 USD
77629-880 77656-148 77643-290 77647-284
1-(2-Ethoxy-3-methylphenyl)cyclobutanol ≥95%
1-(2-Ethoxy-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031805-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR