1-(2-Ethoxy-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

481449-5G 481449-25G 481449-1G 481449-500MG
77633-378EA 1381 USD
77633-378 77659-716 77643-288 77647-282
1-(2-Ethoxy-3-methylphenyl)ethanone ≥95%
1-(2-Ethoxy-3-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
CAS Number: 1226193-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR