2-(2-Ethoxy-3-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

481448-5G 481448-1G 481448-25G 481448-500MG
77629-878EA 1573.84 USD
77629-878 77639-256 77656-146 77651-242
2-(2-Ethoxy-3-methylphenyl)propan-2-ol ≥95%
2-(2-Ethoxy-3-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031789-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR