1-(2-Ethoxy-3-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

481447-5G 481447-1G 481447-500MG
77629-876EA 1489.38 USD
77629-876 77639-254 77647-280
1-(2-Ethoxy-3-methylphenyl)ethanol ≥95%
1-(2-Ethoxy-3-methylphenyl)ethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
CAS Number: 1225606-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR