1-(2-Fluoro-3-methoxypyridin-4-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481444-5G 481444-25G 481444-1G 481444-500MG
77629-874EA 1484.23 USD
77629-874 77656-144 77643-286 77651-240
1-(2-Fluoro-3-methoxypyridin-4-yl)cyclobutanol ≥95%
1-(2-Fluoro-3-methoxypyridin-4-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₀H₁₂FNO₂
Storage Temperature: Ambient
CAS Number: 3031812-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR