2-(2-Fluoro-3-methoxypyridin-4-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

481440-5G 481440-1G 481440-25G 481440-500MG
77629-872EA 1484.23 USD
77629-872 77639-252 77659-712 77647-276
2-(2-Fluoro-3-methoxypyridin-4-yl)propan-2-ol ≥95%
2-(2-Fluoro-3-methoxypyridin-4-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 185.195 g/mol
MW: C₉H₁₂FNO₂
Storage Temperature: Ambient
CAS Number: 3031798-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR