1-(2-Fluoro-5-iodo-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481436-5G 481436-25G 481436-1G 481436-500MG
77629-870EA 1484.23 USD
77629-870 77656-142 77643-284 77647-274
1-(2-Fluoro-5-iodo-3-methylphenyl)cyclobutanol ≥95%
1-(2-Fluoro-5-iodo-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 306.115 g/mol
MW: C₁₁H₁₂FIO
Storage Temperature: Ambient
CAS Number: 3031789-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR