2,2,2-Trifluoro-1-(2-fluoro-5-iodo-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

481435-5G 481435-1G 481435-25G 481435-500MG
77629-868EA 1484.23 USD
77629-868 77639-250 77659-710 77651-238
2,2,2-Trifluoro-1-(2-fluoro-5-iodo-3-methylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-5-iodo-3-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 332.033 g/mol
MW: C₉H₅F₄IO
Storage Temperature: Ambient
CAS Number: 3031789-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR