Ethyl 2-(2-fluoro-5-iodo-3-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

481434-5G 481434-25G 481434-1G 481434-500MG
77629-866EA 1484.23 USD
77629-866 77635-970 77643-282 77647-272
Ethyl 2-(2-fluoro-5-iodo-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-fluoro-5-iodo-3-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 336.098 g/mol
MW: C₁₁H₁₀FIO₃
Storage Temperature: Ambient
CAS Number: 3031798-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR